missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropyl Acetate 95.0+%, TCI America™

Catalog No. B56501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56501G 1 g
B56505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56501G Supplier TCI America Supplier No. B56501G
Only null left

Chemical Identifiers

CAS 592-33-6
Molecular Formula C5H9BrO2
MDL Number MFCD02258489
Synonym Acetic Acid 3-Bromopropyl Ester

Specifications

Boiling Point 163°C
Quantity 1 g
Formula Weight 181.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromopropyl Acetate