missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Bromopropiophenone 98.0+%, TCI America™
Supplier: TCI America B188825G
Specifications
| 3′-Bromopropiophenone | |
| 41°C | |
| 145°C | |
| MFCD00000084 | |
| 3'-bromopropiophenone, 1-3-bromophenyl propan-1-one, 1-propanone, 1-3-bromophenyl, m-bromopropiophenone, 1-3-bromophenyl-1-propanone, 3-bromophenyl ethyl ketone, attercop-chm at106303, pubchem19020, acmc-209f2u | |
| CCC(=O)C1=CC(=CC=C1)Br | |
| 213.074 | |
| 213.07 | |
| Crystalline Powder |
| 19829-31-3 | |
| White-Yellow | |
| C9H9BrO | |
| 25 g | |
| QSHLXVTVXQTHBS-UHFFFAOYSA-N | |
| 1-(3-bromophenyl)propan-1-one | |
| 88272 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 19829-31-3 | |
| 213.074 | |
| QSHLXVTVXQTHBS-UHFFFAOYSA-N | |
| 88272 | |
| CCC(=O)C1=CC(=CC=C1)Br |
| C9H9BrO | |
| MFCD00000084 | |
| 3'-bromopropiophenone, 1-3-bromophenyl propan-1-one, 1-propanone, 1-3-bromophenyl, m-bromopropiophenone, 1-3-bromophenyl-1-propanone, 3-bromophenyl ethyl ketone, attercop-chm at106303, pubchem19020, acmc-209f2u | |
| 1-(3-bromophenyl)propan-1-one |
Safety and Handling
TSCA : Yes