missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromopropiophenone 98.0+%, TCI America™

Catalog No. B18885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18885G 5 g
B188825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18885G Supplier TCI America Supplier No. B18885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19829-31-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD00000084
InChI Key QSHLXVTVXQTHBS-UHFFFAOYSA-N
Synonym 3'-bromopropiophenone, 1-3-bromophenyl propan-1-one, 1-propanone, 1-3-bromophenyl, m-bromopropiophenone, 1-3-bromophenyl-1-propanone, 3-bromophenyl ethyl ketone, attercop-chm at106303, pubchem19020, acmc-209f2u
PubChem CID 88272
IUPAC Name 1-(3-bromophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)Br

Specifications

Melting Point 41°C
Color White-Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 213.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Bromopropiophenone