missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromopropiophenone, 97%

Catalog No. p-7023327
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1675706 5 g
AAA1675714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1675706 Supplier Thermo Scientific Chemicals Supplier No. A1675706
Only null left

CAS: 19829-31-3 | C9H9BrO | 213.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19829-31-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD00000084
InChI Key QSHLXVTVXQTHBS-UHFFFAOYSA-N
Synonym 3'-bromopropiophenone, 1-3-bromophenyl propan-1-one, 1-propanone, 1-3-bromophenyl, m-bromopropiophenone, 1-3-bromophenyl-1-propanone, 3-bromophenyl ethyl ketone, attercop-chm at106303, pubchem19020, acmc-209f2u
PubChem CID 88272
IUPAC Name 1-(3-bromophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)Br

Specifications

Melting Point 38°C to 41°C
Boiling Point 144°C to 145°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1936736
Formula Weight 213.08
Percent Purity 97%
Chemical Name or Material 3'-Bromopropiophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-353-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only