missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionyl Chloride 95.0+%, TCI America™

Catalog No. B206525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B206525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B206525G Supplier TCI America Supplier No. B206525G

Specifications

CAS 15486-96-1
Molecular Formula C3H4BrClO
Quantity 25 g
MDL Number MFCD00000746
UN Number 3265
Synonym 3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2
InChI Key IHBVNSPHKMCPST-UHFFFAOYSA-N
SMILES ClC(=O)CCBr
IUPAC Name 3-bromopropanoyl chloride
Molecular Weight (g/mol) 171.42
PubChem CID 84944
Formula Weight 171.42
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromopropionyl Chloride
Show More Show Less
TSCA Yes
Recommended Storage Refrigerator