Learn More
3-Bromopropionyl chloride, 95%, tech.
CAS: 15486-96-1 | C3H4BrClO | 171.42 g/mol
$161.56 - $161.56
Chemical Identifiers
| CAS | 15486-96-1 |
|---|---|
| Molecular Formula | C3H4BrClO |
| Molecular Weight (g/mol) | 171.42 |
| MDL Number | MFCD00000746 |
| InChI Key | IHBVNSPHKMCPST-UHFFFAOYSA-N |
| Synonym | 3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2 |
| PubChem CID | 84944 |
| IUPAC Name | 3-bromopropanoyl chloride |
| SMILES | ClC(=O)CCBr |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC161990250
|
Thermo Scientific Chemicals
161990250 |
25 g | Glass bottle |
Each for $161.56
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15486-96-1 | |
| 171.42 | |
| IHBVNSPHKMCPST-UHFFFAOYSA-N | |
| 84944 | |
| ClC(=O)CCBr |
| C3H4BrClO | |
| MFCD00000746 | |
| 3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2 | |
| 3-bromopropanoyl chloride |
Specifications
| 15486-96-1 | |
| 100.0 | |
| 1.7000g/mL | |
| 79°C | |
| 94% min. (GC) | |
| C3H4BrClO | |
| BrCH2CH2COCl | |
| 25 g | |
| 1.7 | |
| Solubility in water: reacts | |
| ClC(=O)CCBr | |
| 171.42 | |
| 171.42 | |
| Technical | |
| 3-Bromopropionyl chloride, tech., 95% |
| 94.0 | |
| Yellow | |
| 55°C to 57°C (17.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4920 to 1.497 | |
| MFCD00000746 | |
| 02, II, 231 | |
| 3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2 | |
| IHBVNSPHKMCPST-UHFFFAOYSA-N | |
| 3-bromopropanoyl chloride | |
| 84944 | |
| 95% | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 239-515-2
TSCA : TSCA
RUO – Research Use Only