missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionyl chloride, 95%, tech.

Catalog No. AC161990250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161990250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC161990250 Supplier Thermo Scientific Chemicals Supplier No. 161990250

CAS: 15486-96-1 | C3H4BrClO | 171.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15486-96-1
Molecular Formula C3H4BrClO
Molecular Weight (g/mol) 171.42
MDL Number MFCD00000746
InChI Key IHBVNSPHKMCPST-UHFFFAOYSA-N
Synonym 3-bromopropionyl chloride, propanoyl chloride, 3-bromo, 3-bromopropionylchloride, sgqdjlpqbrmuh@, 3-bromoproionylchloride, 3-bromopropanyl chloride, 3-bromo propionylchloride, 3-bromopropionic chloride, 3-bromopropionoyl chloride, acmc-1bwl2
PubChem CID 84944
IUPAC Name 3-bromopropanoyl chloride
SMILES ClC(=O)CCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 1.7000g/mL
Boiling Point 55°C to 57°C (17.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4920 to 1.497
Linear Formula BrCH2CH2COCl
Quantity 25 g
Beilstein 02, II, 231
Specific Gravity 1.7
Solubility Information Solubility in water: reacts
Formula Weight 171.42
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-Bromopropionyl chloride, tech., 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 239-515-2
TSCA TSCA

RUO – Research Use Only