missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionamide 98.0+%, TCI America™

Catalog No. B14105G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14105G Supplier TCI America Supplier No. B14105G

Chemical Identifiers

CAS 6320-96-3
Molecular Formula C3H6BrNO
Molecular Weight (g/mol) 151.991
MDL Number MFCD00143100
InChI Key DBIVLAVBOICUQX-UHFFFAOYSA-N
PubChem CID 233589
IUPAC Name 3-bromopropanamide
SMILES C(CBr)C(=O)N

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 5 g
Formula Weight 151.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromopropionamide