missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionaldehyde Dimethyl Acetal 98.0+%, TCI America™

Catalog No. b190125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B190125G 25 g
B19015G 5 g
2 options

Catalog No. B190125G

Supplier: TCI America B190125G

Only null left

Chemical Identifiers

CAS 36255-44-4
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.045
MDL Number MFCD00013245
InChI Key ODZZAIFAQLODKN-UHFFFAOYSA-N
Synonym 3-bromopropionaldehyde dimethyl acetal, 1-bromo-3,3-dimethoxypropane, propane, 3-bromo-1,1-dimethoxy, 3-bromo-1,1-dimethoxy-propane, acmc-209ilq, 3,3-dimethoxypropyl bromide, 3-bromopropanal dimethylacetal, ksc495q3l, 3-bromo-1,1dimethoxy-propane, 1,1-dimethoxy-3-bromopropane
PubChem CID 118932
IUPAC Name 3-bromo-1,1-dimethoxypropane
SMILES COC(CCBr)OC

Specifications

Color Yellow
Boiling Point 60°C
Quantity 25 g
UN Number 1993
Formula Weight 183.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromopropionaldehyde Dimethyl Acetal
TSCA No
Recommended Storage Refrigerator