Learn More
3-Bromophenylboronic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 89598-96-9 |
|---|---|
| Molecular Formula | C6H6BBrO2 |
| Molecular Weight (g/mol) | 200.83 |
| MDL Number | MFCD00239386 |
| InChI Key | AFSSVCNPDKKSRR-UHFFFAOYSA-N |
| Synonym | 3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid |
| PubChem CID | 2734318 |
| IUPAC Name | (3-bromophenyl)boronic acid |
| SMILES | B(C1=CC(=CC=C1)Br)(O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC358940010
|
Thermo Scientific Chemicals
358940010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC358940050
|
Thermo Scientific Chemicals
358940050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 89598-96-9 | |
| 200.83 | |
| AFSSVCNPDKKSRR-UHFFFAOYSA-N | |
| 2734318 | |
| B(C1=CC(=CC=C1)Br)(O)O |
| C6H6BBrO2 | |
| MFCD00239386 | |
| 3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid | |
| (3-bromophenyl)boronic acid |
Specifications
| 89598-96-9 | |
| 100.0 | |
| 97% | |
| C6H6BBrO2 | |
| MFCD00239386 | |
| 3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid | |
| AFSSVCNPDKKSRR-UHFFFAOYSA-N | |
| (3-bromophenyl)boronic acid | |
| 2734318 | |
| 97% |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| BrC6H4B(OH)2 | |
| 1g | |
| Solubility in water: 25g/L | |
| B(C1=CC(=CC=C1)Br)(O)O | |
| 200.83 | |
| 200.83 | |
| 3-Bromophenylboronic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
RUO â Research Use Only