missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenethylamine, 99%

CAS: 58971-11-2 | C8H11BrN | 201.09 g/mol

$139.50 - $286.01

Chemical Identifiers

CAS 58971-11-2
Molecular Formula C8H11BrN
Molecular Weight (g/mol) 201.09
MDL Number MFCD01310790
InChI Key ORHRHMLEFQBHND-UHFFFAOYSA-O
Synonym 3-bromophenethylamine, 2-3-bromophenyl ethanamine, 2-3-bromo-phenyl-ethylamine, 2-3-bromophenyl ethylamine, benzeneethanamine, 3-bromo, m-bromophenethylamine, 2-3-bromophenyl ethan-1-amine, 3-bromobenzeneethanamine, 3-bromophenethylaime, 3-bromobenzeneethylamine
PubChem CID 7021736
IUPAC Name 2-(3-bromophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=CC(Br)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350580010
SDS View Documents
Thermo Scientific Chemicals
350580010
1 g Glass Bottle
Each for $139.50
Only null left
 
AC350580050
SDS View Documents
Thermo Scientific Chemicals
350580050
5 g Glass Bottle
Each for $286.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58971-11-2
201.09
ORHRHMLEFQBHND-UHFFFAOYSA-O
7021736
[NH3+]CCC1=CC=CC(Br)=C1
C8H11BrN
MFCD01310790
3-bromophenethylamine, 2-3-bromophenyl ethanamine, 2-3-bromo-phenyl-ethylamine, 2-3-bromophenyl ethylamine, benzeneethanamine, 3-bromo, m-bromophenethylamine, 2-3-bromophenyl ethan-1-amine, 3-bromobenzeneethanamine, 3-bromophenethylaime, 3-bromobenzeneethylamine
2-(3-bromophenyl)ethan-1-aminium

Specifications

58971-11-2
100.0
Authentic
Glass Bottle
1.5735 to 1.5755 (20°C, 589nm)
1 g
3-bromophenethylamine, 2-3-bromophenyl ethanamine, 2-3-bromo-phenyl-ethylamine, 2-3-bromophenyl ethylamine, benzeneethanamine, 3-bromo, m-bromophenethylamine, 2-3-bromophenyl ethan-1-amine, 3-bromobenzeneethanamine, 3-bromophenethylaime, 3-bromobenzeneethylamine
[NH3+]CCC1=CC=CC(Br)=C1
201.09
200.08
Liquid
98.5
Colorless to Yellow
99%
C8H11BrN
MFCD01310790
12, III, 2433
ORHRHMLEFQBHND-UHFFFAOYSA-O
2-(3-bromophenyl)ethan-1-aminium
7021736
99%
3-Bromophenethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only