missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenethyl alcohol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 28229-69-8
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00191857
InChI Key PTTFLKHCSZSFOL-UHFFFAOYSA-N
Synonym 3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol
PubChem CID 2734090
IUPAC Name 2-(3-bromophenyl)ethanol
SMILES OCCC1=CC=CC(Br)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350560010
SDS View Documents
Thermo Scientific Chemicals
350560010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC350560050
SDS View Documents
Thermo Scientific Chemicals
350560050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC350560250
SDS View Documents
Thermo Scientific Chemicals
350560250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

28229-69-8
201.06
PTTFLKHCSZSFOL-UHFFFAOYSA-N
2734090
OCCC1=CC=CC(Br)=C1
C8H9BrO
MFCD00191857
3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol
2-(3-bromophenyl)ethanol

Specifications

28229-69-8
100.0
107°C to 110°C (1.0mmHg)
Authentic
Glass bottle
1.5730 to 1.575
1g
3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol
OCCC1=CC=CC(Br)=C1
201.06
201.06
3-Bromophenethyl alcohol
96.0
1.4780g/mL
>110°C
96% min. (GC)
C8H9BrO
MFCD00191857
1.478
PTTFLKHCSZSFOL-UHFFFAOYSA-N
2-(3-bromophenyl)ethanol
2734090
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only