missing translation for 'onlineSavingsMsg'
Learn More

3-Bromooxetane 95.0+%, TCI America™

Catalog No. B52331G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B52331G Supplier TCI America Supplier No. B52331G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39267-79-3
Molecular Formula C3H5BrO
Molecular Weight (g/mol) 136.98
MDL Number MFCD08544402
InChI Key SZTIZZFKWQWSSP-UHFFFAOYSA-N
Synonym 3-bromo-oxetane, 3-bromo oxetane, 3-bromanyloxetane, 3-bromooxetane, acmc-209j4f, 2-bromo-1,3-epoxypropane, 5-n,n-dimethylamino-2-hydroxybenzaldehyde
PubChem CID 16244493
IUPAC Name 3-bromooxetane
SMILES BrC1COC1

Specifications

Color Yellow
Boiling Point 126°C
Quantity 1 g
UN Number 1993
Formula Weight 136.98
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromooxetane
TSCA No
Recommended Storage Freezer