missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)-3-methyloxetane 97.0+%, TCI America™

Catalog No. B54241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B54241G Supplier TCI America Supplier No. B54241G
Only null left

Chemical Identifiers

CAS 78385-26-9
Molecular Formula C5H9BrO
Molecular Weight (g/mol) 165.03
MDL Number MFCD02684290
InChI Key MGBZKWOJRYGRTO-UHFFFAOYSA-N
Synonym 2-(Bromomethyl)-2-methyl-1,3-epoxypropane
PubChem CID 144860
IUPAC Name 3-(bromomethyl)-3-methyloxetane
SMILES CC1(COC1)CBr

Specifications

Color Yellow
Boiling Point 58°C
Quantity 1 g
Formula Weight 165.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Bromomethyl)-3-methyloxetane