missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuro[3,2-b]pyridine, 96%

Catalog No. AAH3365203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3365203 1 g
AAH3365206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3365203 Supplier Thermo Scientific Chemicals Supplier No. H3365203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 92404-63-2 | C7H4BrNO | 198.019 g/mol

3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 92404-63-2
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD19687763
InChI Key KHMAAFDMZSYVEF-UHFFFAOYSA-N
Synonym 3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
PubChem CID 641477
IUPAC Name 3-bromofuro[3,2-b]pyridine
SMILES C1=CC2=C(C(=CO2)Br)N=C1

Specifications

Melting Point 51°C to 53°C
Quantity 1 g
Formula Weight 198.02
Percent Purity 96%
Chemical Name or Material 3-Bromofuro[3,2-b]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only