missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromofuran 98.0+%, TCI America™
$78.93 - $227.16
Chemical Identifiers
| CAS | 22037-28-1 |
|---|---|
| Molecular Formula | C4H3BrO |
| Molecular Weight (g/mol) | 146.97 |
| MDL Number | MFCD00005347 |
| InChI Key | LXWLEQZDXOQZGW-UHFFFAOYSA-N |
| Synonym | furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz |
| PubChem CID | 89164 |
| IUPAC Name | 3-bromofuran |
| SMILES | BrC1=COC=C1 |
Chemical Identifiers
| 22037-28-1 | |
| 146.97 | |
| LXWLEQZDXOQZGW-UHFFFAOYSA-N | |
| 89164 | |
| BrC1=COC=C1 |
| C4H3BrO | |
| MFCD00005347 | |
| furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz | |
| 3-bromofuran |
Specifications
| 22037-28-1 | |
| C4H3BrO | |
| 1 g | |
| furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz | |
| BrC1=COC=C1 | |
| 146.97 | |
| 146.97 | |
| Liquid |
| 103°C | |
| MFCD00005347 | |
| 1993 | |
| LXWLEQZDXOQZGW-UHFFFAOYSA-N | |
| 3-bromofuran | |
| 89164 | |
| ≥98.0% (GC) | |
| 3-Bromofuran |
Safety and Handling
TSCA : No
Recommended Storage : Freezer