missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuran 98.0+%, TCI America™

Catalog No. B21571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21571G 1 g
B21575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21571G Supplier TCI America Supplier No. B21571G

Chemical Identifiers

CAS 22037-28-1
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES BrC1=COC=C1

Specifications

Boiling Point 103°C
Quantity 1 g
UN Number 1993
Formula Weight 146.97
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromofuran
TSCA No
Recommended Storage Freezer