Learn More
3-Bromofuran, 98%, stabilized with CaCO3
CAS: 22037-28-1 | C4H3BrO | 146.97 g/mol
$277.32 - $277.32
Chemical Identifiers
| CAS | 22037-28-1 |
|---|---|
| Molecular Formula | C4H3BrO |
| Molecular Weight (g/mol) | 146.97 |
| MDL Number | MFCD00005347 |
| InChI Key | LXWLEQZDXOQZGW-UHFFFAOYSA-N |
| Synonym | furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz |
| PubChem CID | 89164 |
| IUPAC Name | 3-bromofuran |
| SMILES | BrC1=COC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC175260025
|
Thermo Scientific Chemicals
175260025 |
2.5 mL | Glass bottle |
Each for $277.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 22037-28-1 | |
| 146.97 | |
| LXWLEQZDXOQZGW-UHFFFAOYSA-N | |
| 89164 | |
| BrC1=COC=C1 |
| C4H3BrO | |
| MFCD00005347 | |
| furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz | |
| 3-bromofuran |
Specifications
| 22037-28-1 | |
| 100.0 | |
| 103.0°C (750.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4960 to 1.4980 | |
| 2.5 mL | |
| furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz | |
| BrC1=COC=C1 | |
| 146.97 | |
| 89164 | |
| 98% | |
| 3-Bromofuran, Stabilized |
| 97.5 | |
| 1.6350g/mL | |
| 3°C | |
| 97.5% min. (GC) | |
| C4H3BrO | |
| MFCD00005347 | |
| 1.635 | |
| LXWLEQZDXOQZGW-UHFFFAOYSA-N | |
| 3-bromofuran | |
| copper | |
| 146.97 | |
| Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 244-744-6
Recommended Storage : −20°C
RUO – Research Use Only