missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuran, 97%, stab. with 0.5% calcium carbonate

CAS: 22037-28-1 | C4H3BrO | 146.97 g/mol

$111.22 - $1,384.00

Chemical Identifiers

CAS 22037-28-1
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES BrC1=COC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2354303
View Documents
Thermo Scientific Chemicals
B2354303
1 g N/A
Only null left
 
AAB2354306
View Documents
Thermo Scientific Chemicals
B2354306
5 g N/A
Only null left
 
AAB2354314
View Documents
Thermo Scientific Chemicals
B2354314
25 g N/A
Only null left
 
Description

Description

3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.

Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
Specifications

Chemical Identifiers

22037-28-1
146.97
LXWLEQZDXOQZGW-UHFFFAOYSA-N
89164
BrC1=COC=C1
C4H3BrO
MFCD00005347
furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
3-bromofuran

Specifications

22037-28-1
102°C to 104°C
C4H3BrO
MFCD00005347
UN1993
furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
LXWLEQZDXOQZGW-UHFFFAOYSA-N
3-bromofuran
89164
97%
1.635
3°C (37°F)
1.496
1 g
105337
Soluble in chloroform,ethyl acetate. Insoluble in water.
BrC1=COC=C1
146.97
146.98
3-Bromofuran, stabilized with 0.5% calcium carbonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 244-744-6

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only