missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuran, 97%, stab. with 0.5% calcium carbonate

Catalog No. AAB2354303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2354303 1 g
AAB2354306 5 g
AAB2354314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2354303 Supplier Thermo Scientific Chemicals Supplier No. B2354303

CAS: 22037-28-1 | C4H3BrO | 146.97 g/mol

3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.

Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.

Chemical Identifiers

CAS 22037-28-1
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES BrC1=COC=C1

Specifications

Density 1.635
Boiling Point 102°C to 104°C
Flash Point 3°C (37°F)
Refractive Index 1.496
Quantity 1 g
UN Number UN1993
Beilstein 105337
Solubility Information Soluble in chloroform,ethyl acetate. Insoluble in water.
Formula Weight 146.98
Percent Purity 97%
Chemical Name or Material 3-Bromofuran, stabilized with 0.5% calcium carbonate
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 244-744-6
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only