missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromofluoranthene 98.0+%, TCI America™
$61.51 - $182.68
Chemical Identifiers
| CAS | 13438-50-1 |
|---|---|
| Molecular Formula | C16H9Br |
| Molecular Weight (g/mol) | 281.15 |
| MDL Number | MFCD07787460 |
| InChI Key | WCXFCLXZMIFHBU-UHFFFAOYSA-N |
| PubChem CID | 13221800 |
| IUPAC Name | 3-bromofluoranthene |
| SMILES | BrC1=CC=C2C3=CC=CC=C3C3=C2C1=CC=C3 |
Chemical Identifiers
| 13438-50-1 | |
| 281.15 | |
| WCXFCLXZMIFHBU-UHFFFAOYSA-N | |
| 3-bromofluoranthene |
| C16H9Br | |
| MFCD07787460 | |
| 13221800 | |
| BrC1=CC=C2C3=CC=CC=C3C3=C2C1=CC=C3 |
Specifications
| 13438-50-1 | |
| 207°C | |
| MFCD07787460 | |
| WCXFCLXZMIFHBU-UHFFFAOYSA-N | |
| 3-bromofluoranthene | |
| 13221800 | |
| ≥98.0% (GC) | |
| 3-Bromofluoranthene |
| 108°C | |
| C16H9Br | |
| 1 g | |
| BrC1=CC=C2C3=CC=CC=C3C3=C2C1=CC=C3 | |
| 281.15 | |
| 281.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No