missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofluoranthene 98.0+%, TCI America™

Catalog No. B45991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45991G 1 g
B45995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45991G Supplier TCI America Supplier No. B45991G
Only null left

Chemical Identifiers

CAS 13438-50-1
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.15
MDL Number MFCD07787460
InChI Key WCXFCLXZMIFHBU-UHFFFAOYSA-N
PubChem CID 13221800
IUPAC Name 3-bromofluoranthene
SMILES BrC1=CC=C2C3=CC=CC=C3C3=C2C1=CC=C3

Specifications

Melting Point 108°C
Boiling Point 207°C
Quantity 1 g
Formula Weight 281.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromofluoranthene