missing translation for 'onlineSavingsMsg'
Learn More

3-Bromochromone 98.0+%, TCI America™

Catalog No. p-7122881
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33345G 5 g
B333425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33345G Supplier TCI America Supplier No. B33345G
Only null left

Chemical Identifiers

CAS 49619-82-1
Molecular Formula C9H5BrO2
Molecular Weight (g/mol) 225.041
MDL Number MFCD00017337
InChI Key IQIGYNPOESZBDJ-UHFFFAOYSA-N
Synonym 3-bromochromone, 3-bromo-4h-chromen-4-one, 3-bromo-4-oxo-4h-1-benzopyran, 3-bromo chromone, maybridge1_006104, acmc-209kh2, 3-bromo-4h-chromen-4-one #, 3-bromo-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,3-bromo
PubChem CID 521256
IUPAC Name 3-bromochromen-4-one
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)Br

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 5 g
Formula Weight 225.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromochromone