missing translation for 'onlineSavingsMsg'
Learn More

3-Bromochlorobenzene, 99%

Catalog No. p-72941
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154200250 25 mL Glass bottle
AC154201000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154200250 Supplier Thermo Scientific Chemicals Supplier No. 154200250
Only null left

CAS: 108-37-2 | C6H4BrCl | 191.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-37-2
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000568
InChI Key JRGGUPZKKTVKOV-UHFFFAOYSA-N
Synonym 3-bromochlorobenzene, m-bromochlorobenzene, 3-chlorobromobenzene, benzene, 1-bromo-3-chloro, m-chlorobromobenzene, 1-chloro-3-bromobenzene, 3-chlorophenyl bromide, m-chlorophenyl bromide, m-bromophenyl chloride, 3-bromo-1-chlorobenzene
PubChem CID 7928
IUPAC Name 1-bromo-3-chlorobenzene
SMILES ClC1=CC=CC(Br)=C1

Specifications

Melting Point -21.5°C
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Density 1.6300g/mL
Quantity 25 mL
Boiling Point 195.0°C to 197.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5760 to 1.5780
Specific Gravity 1.63
Assay Percent Range 98.5% min. (GC)
Linear Formula BrC6H4Cl
Beilstein 05, 209
Solubility Information Solubility in water: insoluble
Formula Weight 191.45
Percent Purity 99%
Chemical Name or Material 3-Bromochlorobenzene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 203-575-8
RTECSNumber CY9005000
TSCA TSCA

RUO – Research Use Only