missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromocarbazole 97.0+%, TCI America™
$109.98 - $384.00
Chemical Identifiers
| CAS | 1592-95-6 |
|---|---|
| Molecular Formula | C12H8BrN |
| Molecular Weight (g/mol) | 246.107 |
| MDL Number | MFCD00222621 |
| InChI Key | LTBWKAYPXIIVPC-UHFFFAOYSA-N |
| Synonym | 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc |
| PubChem CID | 252446 |
| IUPAC Name | 3-bromo-9H-carbazole |
| SMILES | C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br |
Chemical Identifiers
| 1592-95-6 | |
| 246.107 | |
| LTBWKAYPXIIVPC-UHFFFAOYSA-N | |
| 252446 | |
| C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br |
| C12H8BrN | |
| MFCD00222621 | |
| 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc | |
| 3-bromo-9H-carbazole |
Specifications
| 1592-95-6 | |
| White-Yellow | |
| C12H8BrN | |
| 5 g | |
| LTBWKAYPXIIVPC-UHFFFAOYSA-N | |
| 3-bromo-9H-carbazole | |
| 252446 | |
| ≥97.0% (GC) | |
| 3-Bromocarbazole |
| 198°C | |
| 160°C | |
| MFCD00222621 | |
| 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc | |
| C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br | |
| 246.107 | |
| 246.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No