missing translation for 'onlineSavingsMsg'
Learn More

(+)-3-Bromocamphor 98.0+%, TCI America™

Catalog No. B056725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B056725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B056725G Supplier TCI America Supplier No. B056725G
Only null left

Specifications

CAS 10293-06-8
Melting Point 76°C
Color White
Boiling Point 274°C
Molecular Formula C10H15BrO
Quantity 25 g
MDL Number MFCD00003744
Synonym endo-3-bromo-d-camphor, +-camphor bromide, 3s,4s-3-bromo-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one
InChI Key NJQADTYRAYFBJN-MZVBAPTMSA-N
SMILES CC1(C2CCC1(C(=O)C2Br)C)C
IUPAC Name (1S,2S)-2-bromo-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one
Molecular Weight (g/mol) 231.133
PubChem CID 44630109
Formula Weight 231.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-3-Bromocamphor
Show More Show Less
EINECSNumber (4)-0310
RTECSNumber ED6750000
TSCA Yes