missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromoacetyl)pyridine hydrobromide, 98%

Catalog No. AC455520250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455520250 25 g
AC455520050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455520250 Supplier Thermo Scientific Chemicals Supplier No. 455520250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17694-68-7 | C7H7Br2NO | 280.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17694-68-7
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.95
MDL Number MFCD00052182
InChI Key WDTSYONULAZKIE-UHFFFAOYSA-N
Synonym 2-bromo-1-pyridin-3-yl ethanone hydrobromide, 3-bromoacetyl pyridine hydrobromide, 3-2-bromoacetyl pyridine hydrobromide, 2-bromo-1-3-pyridinyl ethanone hydrobromide, 3-bromoacetyl pyridinium bromide, 3-bromoacetyl pyridine hbr, 2-bromo-1-pyridin-3-ylethan-1-one hydrobromide, 2-bromo-1-pyridin-3-ylethanone hydrobromide, 3-bromoacetylpyridine hydrobromide
PubChem CID 2776234
IUPAC Name 2-bromo-1-pyridin-3-ylethanone;hydrobromide
SMILES [H+].[Br-].BrCC(=O)C1=CC=CN=C1

Specifications

Melting Point 192.0°C to 196.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Quantity 25 g
Formula Weight 280.94
Percent Purity 98%
Chemical Name or Material 3-(Bromoacetyl)pyridine hydrobromide
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth
EINECSNumber 241-692-6

RUO – Research Use Only