missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-N,N-dimethylaniline, 97%

Catalog No. AAL1727914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1727914 25 g
AAL1727906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1727914 Supplier Thermo Scientific Chemicals Supplier No. L1727914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16518-62-0 | C8H10BrN | 200.08 g/mol

3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2′,4′,6′-tri-i-propyl-1,1′-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.

Solubility
Miscible with chloroform, dichloromethane and methanol.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 16518-62-0
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00045020
InChI Key USEXQPWLCGBYNT-UHFFFAOYSA-N
Synonym benzenamine, 3-bromo-n,n-dimethyl, 3-bromophenyl dimethylamine, n,n-dimethyl 3-bromoaniline, 3-bromo-n,n-dimethylbenzenamine, n,n-dimethyl-3-bromoaniline, m-bromodimethyl aniline, 3-bromo-dimethylaniline, m-bromodimethylaniline, pubchem3760, acmc-1c80w
PubChem CID 140102
IUPAC Name 3-bromo-N,N-dimethylaniline
SMILES CN(C)C1=CC=CC(Br)=C1

Specifications

Melting Point 11°C
Density 1.365
Boiling Point 126°C (14 mmHg)
Refractive Index 1.6015
Quantity 25 g
Beilstein 2079908
Solubility Information Miscible with chloroform,dichloromethane and methanol.
Formula Weight 200.08
Percent Purity 97%
Chemical Name or Material 3-Bromo-N,N-dimethylaniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only