missing translation for 'onlineSavingsMsg'
Learn More

3-bromo-n-methylaniline, 98%

CAS: 66584-32-5 | C7H8BrN | 186.05 g/mol

$88.48 - $88.48

Chemical Identifiers

CAS 66584-32-5
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD05664376
InChI Key HKOSFZXROYRVJT-UHFFFAOYSA-N
Synonym n-methyl-3-bromoaniline, 3-bromo-n-methyl aniline, benzenamine, 3-bromo-n-methyl, acmc-20an89, 3-bromophenyl-methylamine, 3-bromo-phenyl-methyl-amine, 3-bromo-n-methylaniline
PubChem CID 7018299
IUPAC Name 3-bromo-N-methylaniline
SMILES CNC1=CC(=CC=C1)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC387360010
SDS View Documents
Thermo Scientific Chemicals
387360010
1 g Glass bottle
Each for $88.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

66584-32-5
186.05
HKOSFZXROYRVJT-UHFFFAOYSA-N
7018299
CNC1=CC(=CC=C1)Br
C7H8BrN
MFCD05664376
n-methyl-3-bromoaniline, 3-bromo-n-methyl aniline, benzenamine, 3-bromo-n-methyl, acmc-20an89, 3-bromophenyl-methylamine, 3-bromo-phenyl-methyl-amine, 3-bromo-n-methylaniline
3-bromo-N-methylaniline

Specifications

66584-32-5
100.0
1.4610g/mL
Authentic
Glass bottle
1.6110 to 1.6130 (20°C, 589nm)
MFCD05664376
1.461
HKOSFZXROYRVJT-UHFFFAOYSA-N
3-bromo-N-methylaniline
7018299
98%
3-Bromo-N-methylaniline
97.5
Yellow to Brown
253°C
97.5% min. (GC)
C7H8BrN
BrC6H4NHCH3
1 g
n-methyl-3-bromoaniline, 3-bromo-n-methyl aniline, benzenamine, 3-bromo-n-methyl, acmc-20an89, 3-bromophenyl-methylamine, 3-bromo-phenyl-methyl-amine, 3-bromo-n-methylaniline
CNC1=CC(=CC=C1)Br
186.05
186.05
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only