missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-9-phenylcarbazole 98.0+%, TCI America™
$37.29 - $68.97
Chemical Identifiers
| CAS | 1153-85-1 |
|---|---|
| Molecular Formula | C18H12BrN |
| Molecular Weight (g/mol) | 322.21 |
| MDL Number | MFCD11977305 |
| InChI Key | KUBSCXXKQGDPPD-UHFFFAOYSA-N |
| Synonym | 3-bromo-9-phenyl-9h-carbazole, 3-bromo-n-phenylcarbazole, 9h-carbazole, 3-bromo-9-phenyl, 3-bromo-9-phenyl-carbazole, ksc493s2d, 3-bromanyl-9-phenyl-carbazole, 3-bromo-9-phenyl-9,9a-dihydro-4ah-carbazole |
| PubChem CID | 18942624 |
| IUPAC Name | 3-bromo-9-phenyl-9H-carbazole |
| SMILES | BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 1153-85-1 | |
| 322.21 | |
| KUBSCXXKQGDPPD-UHFFFAOYSA-N | |
| 18942624 | |
| BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 |
| C18H12BrN | |
| MFCD11977305 | |
| 3-bromo-9-phenyl-9h-carbazole, 3-bromo-n-phenylcarbazole, 9h-carbazole, 3-bromo-9-phenyl, 3-bromo-9-phenyl-carbazole, ksc493s2d, 3-bromanyl-9-phenyl-carbazole, 3-bromo-9-phenyl-9,9a-dihydro-4ah-carbazole | |
| 3-bromo-9-phenyl-9H-carbazole |
Specifications
| 1153-85-1 | |
| White | |
| MFCD11977305 | |
| 3-bromo-9-phenyl-9h-carbazole, 3-bromo-n-phenylcarbazole, 9h-carbazole, 3-bromo-9-phenyl, 3-bromo-9-phenyl-carbazole, ksc493s2d, 3-bromanyl-9-phenyl-carbazole, 3-bromo-9-phenyl-9,9a-dihydro-4ah-carbazole | |
| BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1 | |
| 322.21 | |
| 322.21 | |
| Crystalline Powder |
| 79°C | |
| C18H12BrN | |
| 1 g | |
| KUBSCXXKQGDPPD-UHFFFAOYSA-N | |
| 3-bromo-9-phenyl-9H-carbazole | |
| 18942624 | |
| ≥98.0% (GC) | |
| 3-Bromo-9-phenylcarbazole |
Safety and Handling
TSCA : No