missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9-phenylcarbazole 98.0+%, TCI America™

Catalog No. B39081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39081G 1 g
B39085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39081G Supplier TCI America Supplier No. B39081G

Chemical Identifiers

CAS 1153-85-1
Molecular Formula C18H12BrN
Molecular Weight (g/mol) 322.21
MDL Number MFCD11977305
InChI Key KUBSCXXKQGDPPD-UHFFFAOYSA-N
Synonym 3-bromo-9-phenyl-9h-carbazole, 3-bromo-n-phenylcarbazole, 9h-carbazole, 3-bromo-9-phenyl, 3-bromo-9-phenyl-carbazole, ksc493s2d, 3-bromanyl-9-phenyl-carbazole, 3-bromo-9-phenyl-9,9a-dihydro-4ah-carbazole
PubChem CID 18942624
IUPAC Name 3-bromo-9-phenyl-9H-carbazole
SMILES BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 79°C
Color White
Quantity 1 g
Formula Weight 322.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-phenylcarbazole