missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9-phenylcarbazole, 98%

Catalog No. AAH6421206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6421206 5 g
AAH6421214 25 g
AAH6421222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6421206 Supplier Thermo Scientific Chemicals Supplier No. H6421206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1153-85-1 | C18H12BrN | 322.21 g/mol

Used in manufacturing of OLED materials. Also used as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in manufacturing of OLED materials. Also used as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1153-85-1
Molecular Formula C18H12BrN
Molecular Weight (g/mol) 322.21
MDL Number MFCD11977305
InChI Key KUBSCXXKQGDPPD-UHFFFAOYSA-N
Synonym 3-bromo-9-phenyl-9h-carbazole, 3-bromo-n-phenylcarbazole, 9h-carbazole, 3-bromo-9-phenyl, 3-bromo-9-phenyl-carbazole, ksc493s2d, 3-bromanyl-9-phenyl-carbazole, 3-bromo-9-phenyl-9,9a-dihydro-4ah-carbazole
PubChem CID 18942624
IUPAC Name 3-bromo-9-phenylcarbazole
SMILES BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 97°C to 98°C
Quantity 5 g
Beilstein 1315492
Solubility Information Slightly soluble in water.
Formula Weight 322.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Bromo-9-phenylcarbazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only