missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9-(p-tolyl)-9H-carbazole 97.0+%, TCI America™

Catalog No. p-7123921
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50521G 1 g
B50525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50521G Supplier TCI America Supplier No. B50521G
Only null left

Chemical Identifiers

CAS 731016-44-7
Molecular Formula C19H14BrN
Molecular Weight (g/mol) 336.232
MDL Number MFCD16658912
InChI Key RODQKGBYDIAMCF-UHFFFAOYSA-N
PubChem CID 71463937
IUPAC Name 3-bromo-9-(4-methylphenyl)carbazole
SMILES CC1=CC=C(C=C1)N2C3=C(C=C(C=C3)Br)C4=CC=CC=C42

Specifications

Melting Point 158°C
Quantity 1 g
Formula Weight 336.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-(p-tolyl)-9H-carbazole