missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9-(3,5-dimethylphenyl)-9H-carbazole 98.0+%, TCI America™

Catalog No. b5621200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5621200MG 200 mg
B56211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5621200MG Supplier TCI America Supplier No. B5621200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1141017-77-7
Molecular Formula C20H16BrN
Molecular Weight (g/mol) 350.259
InChI Key RSNROIVDIAMOTI-UHFFFAOYSA-N
PubChem CID 131637744
IUPAC Name 3-bromo-9-(3,5-dimethylphenyl)carbazole
SMILES CC1=CC(=CC(=C1)N2C3=C(C=C(C=C3)Br)C4=CC=CC=C42)C

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 200 mg
Formula Weight 350.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-(3,5-dimethylphenyl)-9H-carbazole