missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-9-(2-naphthyl)carbazole 97.0+%, TCI America™
$111.84 - $331.81
Chemical Identifiers
| CAS | 934545-80-9 |
|---|---|
| Molecular Formula | C22H14BrN |
| Molecular Weight (g/mol) | 372.265 |
| MDL Number | MFCD26127422 |
| InChI Key | MBGUQKKYEOAENA-UHFFFAOYSA-N |
| PubChem CID | 68358431 |
| IUPAC Name | 3-bromo-9-naphthalen-2-ylcarbazole |
| SMILES | C1=CC=C2C=C(C=CC2=C1)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 |
Chemical Identifiers
| 934545-80-9 | |
| 372.265 | |
| MBGUQKKYEOAENA-UHFFFAOYSA-N | |
| 3-bromo-9-naphthalen-2-ylcarbazole |
| C22H14BrN | |
| MFCD26127422 | |
| 68358431 | |
| C1=CC=C2C=C(C=CC2=C1)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 |
Specifications
| 934545-80-9 | |
| White-Yellow | |
| MFCD26127422 | |
| MBGUQKKYEOAENA-UHFFFAOYSA-N | |
| 3-bromo-9-naphthalen-2-ylcarbazole | |
| 68358431 | |
| ≥97.0% (HPLC,N) | |
| 3-Bromo-9-(2-naphthyl)carbazole |
| 133°C | |
| C22H14BrN | |
| 1 g | |
| C1=CC=C2C=C(C=CC2=C1)N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53 | |
| 372.265 | |
| 372.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No