missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9-(1-naphthyl)-9H-carbazole 97.0+%, TCI America™

Numéro de catalogue. B5050200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B5050200MG 200 mg
B50501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B5050200MG Fournisseur TCI America Code fournisseur B5050200MG
Il en reste null

Identifiants chimiques

CAS 934545-83-2
Molecular Formula C22H14BrN
Molecular Weight (g/mol) 372.265
MDL Number MFCD26127423
InChI Key QTWVKYHIDBPQIL-UHFFFAOYSA-N
PubChem CID 59561149
IUPAC Name 3-bromo-9-naphthalen-1-ylcarbazole
SMILES C1=CC=C2C(=C1)C=CC=C2N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53

Spécifications

Color White-Yellow
Quantity 200 mg
Formula Weight 372.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-(1-naphthyl)-9H-carbazole