missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-7-nitroindazole, 98+%

Catalog No. AAJ6348006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6348006 5 g
AAJ6348003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6348006 Supplier Thermo Scientific Chemicals Supplier No. J6348006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of nitric oxide synthase (NOS) | CAS: 74209-34-0 | C7H4BrN3O2 | 242.03 g/mol

Inhibitor of nitric oxide synthase (NOS)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74209-34-0
Molecular Formula C7H4BrN3O2
Molecular Weight (g/mol) 242.03
MDL Number MFCD00159910
InChI Key NFSTZPMYAZRZPC-UHFFFAOYSA-N
Synonym 3-bromo-7-nitroindazole, 3-bromo-7-nitro-1h-indazole, 1h-indazole,3-bromo-7-nitro, brni, 3-bromo-7ni, 3-br-7-nitroindazole, tocris-0735, lopac-b-2050, d0u6zt, d0x5qj
PubChem CID 1649
IUPAC Name 3-bromo-7-nitro-2H-indazole
SMILES [O-][N+](=O)C1=CC=CC2=C(Br)NN=C12

Specifications

Color Yellow
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Soluble in ethanol at 25mg/ml; Also soluble in DMSO
Formula Weight 242
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 3-Bromo-7-nitroindazole
Hazard Category H360
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only