missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-6-methoxy-2-methylpyridine 98.0+%, TCI America™
$39.14 - $93.21
Chemical Identifiers
| CAS | 126717-59-7 |
|---|---|
| Molecular Formula | C7H8BrNO |
| Molecular Weight (g/mol) | 202.051 |
| MDL Number | MFCD03094943 |
| InChI Key | VWNXCCTWVAQAPL-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl |
| PubChem CID | 10798146 |
| IUPAC Name | 3-bromo-6-methoxy-2-methylpyridine |
| SMILES | CC1=C(C=CC(=N1)OC)Br |
Chemical Identifiers
| 126717-59-7 | |
| 202.051 | |
| VWNXCCTWVAQAPL-UHFFFAOYSA-N | |
| 10798146 | |
| CC1=C(C=CC(=N1)OC)Br |
| C7H8BrNO | |
| MFCD03094943 | |
| 5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl | |
| 3-bromo-6-methoxy-2-methylpyridine |
Specifications
| 126717-59-7 | |
| 86°C | |
| MFCD03094943 | |
| 5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl | |
| CC1=C(C=CC(=N1)OC)Br | |
| 202.051 | |
| 202.05 | |
| Liquid |
| Yellow | |
| C7H8BrNO | |
| 1 g | |
| VWNXCCTWVAQAPL-UHFFFAOYSA-N | |
| 3-bromo-6-methoxy-2-methylpyridine | |
| 10798146 | |
| ≥98.0% (GC) | |
| 3-Bromo-6-methoxy-2-methylpyridine |
Safety and Handling
TSCA : No