missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-methoxy-2-methylpyridine 98.0+%, TCI America™

Catalog No. B44605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44605G 5 g
B44601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44605G Supplier TCI America Supplier No. B44605G
Only null left

Chemical Identifiers

CAS 126717-59-7
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD03094943
InChI Key VWNXCCTWVAQAPL-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl
PubChem CID 10798146
IUPAC Name 3-bromo-6-methoxy-2-methylpyridine
SMILES CC1=C(C=CC(=N1)OC)Br

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
Formula Weight 202.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-6-methoxy-2-methylpyridine