missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-fluoro-2-methylpyridine 98.0+%, TCI America™

Catalog No. p-7123978
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51355G 5 g
B513525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51355G Supplier TCI America Supplier No. B51355G

Chemical Identifiers

CAS 375368-83-5
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.02
MDL Number MFCD03095092
InChI Key GUYGQQWIKZLHTP-UHFFFAOYSA-N
Synonym 3-Bromo-6-fluoro-2-picoline
PubChem CID 17750149
IUPAC Name 3-bromo-6-fluoro-2-methylpyridine
SMILES CC1=C(Br)C=CC(F)=N1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-6-fluoro-2-methylpyridine