missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals

Catalog No. p-7032921
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3555803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3555803 Supplier Thermo Scientific Chemicals Supplier No. H3555803

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 886371-28-4
Molecular Formula C7H4BrClN2
Molecular Weight (g/mol) 231.477
MDL Number MFCD08667858
InChI Key LIBKDAUSMLPFBF-UHFFFAOYSA-N
Synonym 3-bromo-6-chloroimidazo 1,2-a pyridine, 6-chloro-3-bromo-imidazo 1,2-a pyridine, acmc-209qvk, 3-bromo-6-chloro-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 3-bromo-6-chloro
PubChem CID 45790042
IUPAC Name 3-bromo-6-chloroimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=C(N2C=C1Cl)Br

Specifications

Melting Point 105°C to 107°C
Quantity 1 g
Formula Weight 231.48
Percent Purity 95%
Chemical Name or Material 3-Bromo-6-chloroimidazo[1,2-a]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only