missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-chlorochromone 98.0+%, TCI America™

Catalog No. B49671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49671G 1 g
B49675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49671G Supplier TCI America Supplier No. B49671G

Chemical Identifiers

CAS 73220-38-9
Molecular Formula C9H4BrClO2
Molecular Weight (g/mol) 259.48
MDL Number MFCD02093123
InChI Key ZILSBPAUJJBEFF-UHFFFAOYSA-N
Synonym 3-bromo-6-chloro-4h-chromen-4-one, 3-bromo-6-chlorochromone, 4h-1-benzopyran-4-one,3-bromo-6-chloro, 3-bromo-6-chloro chromone, 3-bromo-6-chloro-4-oxo-4h-1-benzopyran, 3-bromo-6-chloro-4h-1-benzopyran-4-one
PubChem CID 3498945
IUPAC Name 3-bromo-6-chloro-4H-chromen-4-one
SMILES ClC1=CC=C2OC=C(Br)C(=O)C2=C1

Specifications

Melting Point 130°C
Quantity 1 g
Formula Weight 259.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-6-chlorochromone