missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)pyridine, 95%

Catalog No. AAH2582303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2582303 1 g
AAH2582306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2582303 Supplier Thermo Scientific Chemicals Supplier No. H2582303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 436799-33-6 | C6H3BrF3N | 225.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 436799-33-6
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD04972700
InChI Key HEDHNDVPKRVQPN-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 5-bromo-3-trifluoromethyl pyridine, pyridine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluormethyl-pyridine, abbypharma ap-11-5533, 3-trifluoromethyl-5-bromopyridine, pubchem3010, acmc-209jv5, 3-bromo-5-trifluoromethylpyridin
PubChem CID 11127991
IUPAC Name 3-bromo-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC=C1Br)C(F)(F)F

Specifications

Melting Point 44°C to 46°C
Boiling Point 65°C to 66°C (19 mmHg)
Flash Point 85°C (185°F)
Quantity 1 g
Formula Weight 226
Percent Purity 95%
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H335-H500
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only