missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)benzene-1,2-diamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB05134DA

Quantity 1g

 View more versions of this product

Catalog No. BTB05134DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

113170-72-2
255.038
OVGAPUVMLZWQQF-UHFFFAOYSA-N
735785
C1=C(C=C(C(=C1N)N)Br)C(F)(F)F
C7H6BrF3N2
MFCD00042484
3-bromo-5-trifluoromethyl benzene-1,2-diamine, 3-bromo-4,5-diaminobenzotrifluoride, buttpark 82\03-57, 1,2-diamino-3-bromo-5-trifluoromethyl benzene, 3-bromo-5-trifluoromethyl-1,2-phenylenediamine, 2-amino-3-bromo-5-trifluoromethyl phenylamine, 3-bromo-5-trifluoromethyl-1,2-benzenediamine, 4,5-diamino-3-bromobenzotrifluoride, pubchem2253
3-bromo-5-(trifluoromethyl)benzene-1,2-diamine

Specifications

3-Bromo-5-(trifluoromethyl)benzene-1, 2-diamine
96.5
97%
C7H6BrF3N2
MFCD00042484
OVGAPUVMLZWQQF-UHFFFAOYSA-N
3-bromo-5-(trifluoromethyl)benzene-1,2-diamine
735785
97%
113170-72-2
100.0
Amber glass bottle
1g
3-bromo-5-trifluoromethyl benzene-1,2-diamine, 3-bromo-4,5-diaminobenzotrifluoride, buttpark 82\03-57, 1,2-diamino-3-bromo-5-trifluoromethyl benzene, 3-bromo-5-trifluoromethyl-1,2-phenylenediamine, 2-amino-3-bromo-5-trifluoromethyl phenylamine, 3-bromo-5-trifluoromethyl-1,2-benzenediamine, 4,5-diamino-3-bromobenzotrifluoride, pubchem2253
C1=C(C=C(C(=C1N)N)Br)C(F)(F)F
255.038
255.04
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents