missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methylpyridine 98.0+%, TCI America™

Catalog No. B37245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37245G 5 g
B37241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37245G Supplier TCI America Supplier No. B37245G
Only null left

Chemical Identifiers

CAS 3430-16-8
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD01646141
InChI Key ADCLTLQMVAEBLB-UHFFFAOYSA-N
Synonym 5-bromo-3-picoline, 3-bromo-5-picoline, 3-bromo-5-methyl-pyridine, 3-methyl-5-bromopyridine, pyridine, 3-bromo-5-methyl, 5-bromo-3-methylpyridine, 3-brom-5-methylpyridin, zlchem 381, 5-bromo-3-picolin, pubchem4036
PubChem CID 817713
IUPAC Name 3-bromo-5-methylpyridine
SMILES CC1=CC(Br)=CN=C1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-methylpyridine