missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methoxypyridine 97.0+%, TCI America™

Catalog No. B36845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36845G 5 g
B36841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36845G Supplier TCI America Supplier No. B36845G

Chemical Identifiers

CAS 50720-12-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD00234169
InChI Key FZWUIWQMJFAWJW-UHFFFAOYSA-N
Synonym 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085
PubChem CID 817163
IUPAC Name 3-bromo-5-methoxypyridine
SMILES COC1=CC(=CN=C1)Br

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 188.02
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Bromo-5-methoxypyridine