Learn More
3-Bromo-5-methoxypyridine, 97%, Thermo Scientific Chemicals
$187.12 - $187.12
Chemical Identifiers
| CAS | 50720-12-2 |
|---|---|
| Molecular Formula | C6H6BrNO |
| Molecular Weight (g/mol) | 188.024 |
| MDL Number | MFCD00234169 |
| InChI Key | FZWUIWQMJFAWJW-UHFFFAOYSA-N |
| Synonym | 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085 |
| PubChem CID | 817163 |
| IUPAC Name | 3-bromo-5-methoxypyridine |
| SMILES | COC1=CC(=CN=C1)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2772806
|
Thermo Scientific Chemicals
H2772806 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 50720-12-2 | |
| 188.024 | |
| FZWUIWQMJFAWJW-UHFFFAOYSA-N | |
| 817163 | |
| COC1=CC(=CN=C1)Br |
| C6H6BrNO | |
| MFCD00234169 | |
| 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085 | |
| 3-bromo-5-methoxypyridine |
Specifications
| 50720-12-2 | |
| 102°C (216°F) | |
| MFCD00234169 | |
| 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085 | |
| COC1=CC(=CN=C1)Br | |
| 188.024 | |
| 188.02 | |
| 3-Bromo-5-methoxypyridine |
| 29°C to 33°C | |
| C6H6BrNO | |
| 5 g | |
| FZWUIWQMJFAWJW-UHFFFAOYSA-N | |
| 3-bromo-5-methoxypyridine | |
| 817163 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only