missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methoxypyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH2772806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2772806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2772806 Supplier Thermo Scientific Chemicals Supplier No. H2772806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50720-12-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD00234169
InChI Key FZWUIWQMJFAWJW-UHFFFAOYSA-N
Synonym 3-bromo-5-methoxy-pyridine, 5-methoxy-3-bromopyridine, 5-bromo-3-methoxypyridine, pyridine, 3-bromo-5-methoxy, 5-bromopyridin-3-yl methyl ether, 3-methoxy-5-bromopyridine, 3-bromo-5-methoxy pyridine, 3-brom-5-methoxypyridin, zlchem 930, pubchem5085
PubChem CID 817163
IUPAC Name 3-bromo-5-methoxypyridine
SMILES COC1=CC(=CN=C1)Br

Specifications

Melting Point 29°C to 33°C
Flash Point 102°C (216°F)
Quantity 5 g
Formula Weight 188.02
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-methoxypyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only