missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromo-5'-fluoroacetophenone, 96%

Catalog No. AAH6297703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6297703 1 g
AAH6297706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6297703 Supplier Thermo Scientific Chemicals Supplier No. H6297703

CAS: 105515-20-6 | C8H6BrFO | 217.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105515-20-6
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD11847746
InChI Key MZDXPUDHZBCYGS-UHFFFAOYSA-N
Synonym 1-3-bromo-5-fluorophenyl ethanone, 3'-bromo-5'-fluoroacetophenone, 1-3-bromo-5-fluorophenyl ethan-1-one, ethanone, 1-3-bromo-5-fluorophenyl, 3-bromo-5-fluoroacetophenone, pubchem22227, 1-3-bromo-5-fluoro-phenyl-ethanone
PubChem CID 13730359
IUPAC Name 1-(3-bromo-5-fluorophenyl)ethanone
SMILES CC(=O)C1=CC(F)=CC(Br)=C1

Specifications

Quantity 1 g
Formula Weight 217.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3'-Bromo-5'-fluoroacetophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only