missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-fluoro-2-cyanopyridine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 950670-18-5 |
|---|---|
| Molecular Formula | C6H2BrFN2 |
| Molecular Weight (g/mol) | 201 |
| InChI Key | OLOSREGULYFXDD-UHFFFAOYSA-N |
| Synonym | 3-bromo-5-fluoropicolinonitrile, 3-bromo-5-fluoro-2-cyanopyridine, 3-bromo-2-cyano-5-fluoropyridine, 3-bromo-5-fluoro-2-pyridinecarbonitrile, abbypharma ap-18-5887, 3-aromo-5-fluoropicolinonitrile |
| PubChem CID | 46941477 |
| IUPAC Name | 3-bromo-5-fluoropyridine-2-carbonitrile |
| SMILES | C1=C(C=NC(=C1Br)C#N)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC451430010
|
Thermo Scientific Chemicals
451430010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC451430050
|
Thermo Scientific Chemicals
451430050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 950670-18-5 | |
| 201 | |
| 3-bromo-5-fluoropicolinonitrile, 3-bromo-5-fluoro-2-cyanopyridine, 3-bromo-2-cyano-5-fluoropyridine, 3-bromo-5-fluoro-2-pyridinecarbonitrile, abbypharma ap-18-5887, 3-aromo-5-fluoropicolinonitrile | |
| 3-bromo-5-fluoropyridine-2-carbonitrile |
| C6H2BrFN2 | |
| OLOSREGULYFXDD-UHFFFAOYSA-N | |
| 46941477 | |
| C1=C(C=NC(=C1Br)C#N)F |
Specifications
| 950670-18-5 | |
| Glass Bottle | |
| 1g | |
| OLOSREGULYFXDD-UHFFFAOYSA-N | |
| 3-bromo-5-fluoropyridine-2-carbonitrile | |
| 46941477 | |
| 97% |
| 96% min. (GC) | |
| C6H2BrFN2 | |
| 3-bromo-5-fluoropicolinonitrile, 3-bromo-5-fluoro-2-cyanopyridine, 3-bromo-2-cyano-5-fluoropyridine, 3-bromo-5-fluoro-2-pyridinecarbonitrile, abbypharma ap-18-5887, 3-aromo-5-fluoropicolinonitrile | |
| C1=C(C=NC(=C1Br)C#N)F | |
| 201 | |
| 201 | |
| 3-Bromo-5-fluoro-2-cyanopyridine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS Signal Word: Warning
RUO â Research Use Only